VWR® Basic Visitor Coats, Velcro® Closure
White, S, 96 cm
Catalog # 113-8956
Supplier: VWR Collection
Jump to:
Product Details & Documents
These disposable coats, with collar, are made from non woven, spunbonded polypropylene (SPP) fabric, which protects against dirt, grime, and certain dry particulates, in non hazardous environments.
- VELCRO® closure
- Elastic wrists
- Free from natural rubber latex
- Available in white and blue
Weight: 30 g/m²
: 1 piece per bag, 50 bags per case.
Documents
Specifications
- Clothing Size:S
- Colour:White
- Length:96 cm
- Seq:02
- Cat. No.:113-8956
- Garment application:Laboratory
- Gender:Unisex
- Pockets:Without pockets
- Closure type:Hook-and-loop / VELCRO®
- Weight:30 g/m²



