VWR® Basic Visitor Coats, Velcro® Closure
White, XS, 92 cm
Catalog # 113-8955
Supplier: VWR Collection
Jump to:
Product Details & Documents
These disposable coats, with collar, are made from non woven, spunbonded polypropylene (SPP) fabric, which protects against dirt, grime, and certain dry particulates, in non hazardous environments.
- VELCRO® closure
- Elastic wrists
- Free from natural rubber latex
- Available in white and blue
Weight: 30 g/m²
Packaging: 1 piece per bag, 50 bags per case.
Documents
Specifications
- Clothing Size:XS
- Colour:White
- Length:92 cm
- Cat. No.:113-8955
- Garment application:Laboratory
- Gender:Unisex
- Pockets:Without pockets
- Closure type:Hook-and-loop / VELCRO®
- Weight:30 g/m²
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



