VWR® Basic, Bouffant Caps, Latex-Free
Yellow, 580 mm
Catalog # 113-8882
Supplier: VWR Collection
Jump to:
Product Details & Documents
These disposable caps are made from a non woven, spunbonded polypropylene (SPP) fabric, which protects against dirt, grime, and certain dry particulates, in non hazardous environments.
- Free from natural rubber latex
- Available in a wide range of colours
Weight: 10 g/m²
Packaging: 100 pieces per bag, 10 bags per case.
Documents
CoC VWR® Basic Bouffant Caps.pdf
Conformity declaration
CoC_VWRI113-1459_03200637 COC_VWRI113-1459_03221002 COC_VWRI113-1459_03221003 COC_VWRI113-1459_03231100 COC_VWRI113-1459_03231364
More
Food Contact Certificate
Additional Product Details
VWR® Non Regulated Disposables for Production
Product Literature
Specifications
- Ø:580 mm
- Colour:Yellow
- Seq:24
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



