VWR® Basic, Bouffant Caps, Latex-Free
Red, 610 mm
Catalog # 113-8886
Supplier: VWR Collection
Jump to:
Product Details & Documents
These disposable caps are made from a non woven, spunbonded polypropylene (SPP) fabric, which protects against dirt, grime, and certain dry particulates, in non hazardous environments.
- Free from natural rubber latex
- Available in a wide range of colours
Weight: 10 g/m²
Packaging: 100 pieces per bag, 10 bags per case.
Specifications
- Ø:610 mm
- Colour:Red
- Seq:14



