ContactUsLinkComponent
Human EGF R, Fc Tag (EGR-H5252) is expressed from human 293 cells (HEK293). It contains AA Leu 25 - Ser 645 (Accession P00533-1). 1 * 1 mg
Catalog # ACRBEGR-H5252-1MG
Supplier:  ACRO Biosystems
Human EGF R, Fc Tag (EGR-H5252) is expressed from human 293 cells (HEK293). It contains AA Leu 25 - Ser 645 (Accession P00533-1). 1 * 1 mg
Catalog # ACRBEGR-H5252-1MG
Supplier:  ACRO Biosystems
Frequently Bought Together