Order Entry
Germany
Orders LinkContactUsLinkComponent
3,3'-BIS(1,1,1,3,3,3-HEXAFLUORO-2-HYDROXY-2-PROPYL)-5 ,5'-DIMETHYLBENZIDINE[HFA-MTB

3,3'-BIS(1,1,1,3,3,3-HEXAFLUORO-2-HYDROXY-2-PROPYL)-5 ,5'-DIMETHYLBENZIDINE[HFA-MTB

Catalog # XXXXHFA-MTB
Supplier:   Chemicals
3,3'-BIS(1,1,1,3,3,3-HEXAFLUORO-2-HYDROXY-2-PROPYL)-5 ,5'-DIMETHYLBENZIDINE[HFA-MTB
Catalog # XXXXHFA-MTB
Supplier:  Chemicals
CAS Number:  
3,3'-BIS(1,1,1,3,3,3-HEXAFLUORO-2-HYDROXY-2-PROPYL)-5 ,5'-DIMETHYLBENZIDINE[HFA-MTB
CAS Number:  
Price
2 330,00 €
Log in for availability
1 * 1 kg
Product Details & Documents
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.