Order Entry
Germany
Orders LinkContactUsLinkComponent
VWR® Premium Plus, Slide Boxes

VWR® Premium Plus, Slide Boxes

Slide boxes, Premium Plus, Green, PP, 100, 195×207×32 mm
Catalog # 631-1244
Supplier:   VWR Collection
VWR® Premium Plus, Slide Boxes
Catalog # 631-1244
Supplier:  VWR Collection
VWR® Premium Plus, Slide Boxes
VWR® Premium Plus, Slide Boxes

Slide boxes, Premium Plus, Green, PP, 100, 195×207×32 mm

Price
97,10 €
Log in for availability
1 * 5 ST
Product Details & Documents

These slide boxes feature multiple labelling areas, and an index card can be used as the lid is transparent.

  • Slide rail design ensures a secure fit
  • Central numbering grid for slide organisation and easy viewing
  • Plastic design eliminates potential rusting of metal parts
  • Sliding lock provides secure storage
  • Hinged lid smoothly opens 180°, sits flat on surface preventing misplacement
  • EVA foam lines the bottom of the box
Specifications
  • No. of slides:
    100
  • Colour:
    Green
  • Description:
    Slide boxes, Premium Plus
  • Material:
    PP
  • W×D×H:
    195×207×32 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.