Order Entry
Germany
Orders LinkContactUsLinkComponent
 

Dosing spoons, SteriWare®

Sterile, 2,5/5,0 ml
Catalog # SAMP8642A-01S
Supplier:   Sampling Systems
Dosing spoons, SteriWare®
Catalog # SAMP8642A-01S
Supplier:  Sampling Systems
Dosing spoons, SteriWare®
Dosing spoons, SteriWare®

Sterile, 2,5/5,0 ml

Price
79,90 €
Log in for availability
1 * 130 ST
Product Details & Documents

These disposable and lightweight PS spoons are ideal for sampling and dosing small amounts of powder and liquid. Ideal for work in quality control, laboratories and production.

  • Made from white shatterproof high impact polystyrene (FDA and EU 10/2011 acceptable grade)
  • Handy design comes with a 2,5 ml and 5 ml spoon on either end
  • Available in both sterile and non sterile versions
  • Cross contamination is eliminated - the device can be used straight from the bag
  • Moulded and packed in a cleanroom to assure cleanliness and hygiene
  • BSE/TSE-free

 : Individually bagged and shipped with full documentation including certificates of conformity and sterilisation certificates.

Specifications
  • Capacity:
    2,5/5,0 ml
  • Description:
    Sterile
  • Seq:
    02