Order Entry
Germany
Orders LinkContactUsLinkComponent
Dosing spoons, SteriWare®

Dosing spoons, SteriWare®

Non sterile, 2,5/5,0 ml
Catalog # SAMP8642A-01
Supplier:   Sampling Systems
Dosing spoons, SteriWare®
Catalog # SAMP8642A-01
Supplier:  Sampling Systems
Dosing spoons, SteriWare®
Dosing spoons, SteriWare®

Non sterile, 2,5/5,0 ml

Price
57,30 €
Log in for availability
1 * 130 ST
Product Details & Documents

These disposable and lightweight PS spoons are ideal for sampling and dosing small amounts of powder and liquid. Ideal for work in quality control, laboratories and production.

  • Made from white shatterproof high impact polystyrene (FDA and EU 10/2011 acceptable grade)
  • Handy design comes with a 2,5 ml and 5 ml spoon on either end
  • Available in both sterile and non sterile versions
  • Cross contamination is eliminated - the device can be used straight from the bag
  • Moulded and packed in a cleanroom to assure cleanliness and hygiene
  • BSE/TSE-free

Packaging: Individually bagged and shipped with full documentation including certificates of conformity and sterilisation certificates.

Specifications
  • Capacity:
    2,5/5,0 ml
  • Description:
    Non sterile
  • Seq:
    01
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.