ContactUsLinkComponent
Sensor type: InPro6800 Shank diameter: 12mm Shaft material: 1.4404 [1] A-length in mm: 70mm (d12) Version Process Adaptation: Threaded sleeve Pg13.5 [1] Material Wetted: 1.4404 [1] Execution Schafthuelse: Standard [1] Membrane type: Teflon [1] Electr 1 * 1 items
Catalog # METN52200964
Supplier:  Mettler - Toledo
Sensor type: InPro6800 Shank diameter: 12mm Shaft material: 1.4404 [1] A-length in mm: 70mm (d12) Version Process Adaptation: Threaded sleeve Pg13.5 [1] Material Wetted: 1.4404 [1] Execution Schafthuelse: Standard [1] Membrane type: Teflon [1] Electr 1 * 1 items
Catalog # METN52200964
Supplier:  Mettler - Toledo
Frequently Bought Together