ContactUsLinkComponent
ECO SERIES GLASS COLUMN, AB-VERSION, TYPE VARIO , 25 [MM] ID, 15 [BAR] PRESSURE LIMIT, 0 - 120 [MM] BED LENGTH, 0 - 60 [ML] VOLUME, 16 - 40 [µM] FRIT POROSITY 1 * 1 ST
Catalog # YMCAECO25/120V3V
Supplier:  YMC
ECO SERIES GLASS COLUMN, AB-VERSION, TYPE VARIO , 25 [MM] ID, 15 [BAR] PRESSURE LIMIT, 0 - 120 [MM] BED LENGTH, 0 - 60 [ML] VOLUME, 16 - 40 [µM] FRIT POROSITY 1 * 1 ST
Catalog # YMCAECO25/120V3V
Supplier:  YMC
Frequently Bought Together