Digital Solutions
Applications
Support
About
Germany
ContactUsLinkComponent
GPC COLUMN PSS PROTEEMA MICRO IN WATER MOLAR MASS RANGE 1 000 - 1 200 000 DA 1 * 1 ST
POSSPSM0525033E2
:
POLYMER STANDARDS SERVICE
GPC COLUMN PSS PROTEEMA MICRO IN WATER MOLAR MASS RANGE 1 000 - 1 200 000 DA 1 * 1 ST
POSSPSM0525033E2
:
POLYMER STANDARDS SERVICE