Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
GPC COLUMN PSS POLARSIL MICRO IN DMF MOLAR MASS RANGE 100 - 100 000 DA 1 * 1 ST
Catalog # POSSPSM0525031E2
Supplier:
POLYMER STANDARDS SERVICE
Expand image
GPC COLUMN PSS POLARSIL MICRO IN DMF MOLAR MASS RANGE 100 - 100 000 DA 1 * 1 ST
Catalog # POSSPSM0525031E2
Supplier:
POLYMER STANDARDS SERVICE