Order Entry
Germany
Orders LinkContactUsLinkComponent
Human recombinant PEDF

Human recombinant PEDF

5 µG
Catalog # APOSBITP2095-5UG
Supplier:   Apollo Scientific
Human recombinant PEDF
Catalog # APOSBITP2095-5UG
Supplier:  Apollo Scientific
CAS Number:  
Human recombinant PEDF

5 µG

CAS Number:  
Price
160,00 €
Log in for availability
1 * 5 µG
Product Details & Documents
Specifications

Specifications

  • Pk:
    5 µG
  • Cat. No.:
    APOSBITP2095-5UG
  • Protein/peptide type:
    Recombinant
  • Protein/peptide name:
    PEDF
  • Protein synonyms:
    Pigment Epithelium-Derived Factor
  • Species:
    Human

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.