Methylisonicotinate-N-oxide
Bottle, 25 g
Catalog # APOSOR13135-25G
Supplier: Apollo Scientific
Methylisonicotinate-N-oxide
Catalog # APOSOR13135-25G
Supplier: Apollo Scientific
CAS Number: 3783-38-8
Jump to:
Product Details & Documents
Synonyms: 4-(Methoxycarbonyl)pyridine 1-oxide
- CAS Number:3783-38-8
- Formula:C₇H₇NO₃
- MDL Number:MFCD00128856
- Molecular Weight:153.14 g/mol
- Melting Pt:121…123 °C
- Storage Temperature:Ambient
Specifications
Specifications
- Pack type:Bottle
- Pk:25 g
- Cat. No.:APOSOR13135-25G
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



