Order Entry
Germany
Orders LinkContactUsLinkComponent
2-(4-Bromophenyl)ethanol

2-(4-Bromophenyl)ethanol

Bottle, 5 g
Catalog # APOSOR30084-5G
Irritant
Supplier:   Apollo Scientific
2-(4-Bromophenyl)ethanol
Catalog # APOSOR30084-5G
Supplier:  Apollo Scientific
CAS Number:  4654-39-1
2-(4-Bromophenyl)ethanol

Bottle, 5 g

CAS Number:  4654-39-1
Price
29,00 €
Log in for availability
1 * 5 g
Product Details & Documents
Synonyms:  4-Bromophenethyl alcohol
  • CAS Number:4654-39-1
  • Formula:C₈H₉BrO
  • MDL Number:MFCD00002897
  • Molecular Weight:201.06 g/mol
  • Boiling Pt:138 °C/9mmHg
  • Density:1.436 g/cm³ (25 °C)
  • Flash Pt:110 °C
  • Storage Temperature:Ambient
  • EINECS:225-093-7
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g

Specifications