2-(4-Bromophenyl)ethanol
Bottle, 25 g
APOSOR30084-25G
: 4-Bromophenethyl alcohol
- :4654-39-1
- :C₈H₉BrO
- :MFCD00002897
- :201.06 g/mol
- :138 °C/9mmHg
- :1.436 g/cm³ (25 °C)
- :110 °C
- :Ambient
- :225-093-7
- Pack type:Bottle
- Pk:25 g