Order Entry
Germany
Orders LinkContactUsLinkComponent
 

2-Bromothiazole

Bottle, 25 g
Catalog # APOSOR23133-25G
Irritant
Supplier:   Apollo Scientific
2-Bromothiazole
Catalog # APOSOR23133-25G
Supplier:  Apollo Scientific
CAS Number:  3034-53-5
2-Bromothiazole

Bottle, 25 g

CAS Number:  3034-53-5
Price
21,80 €
Log in for availability
1 * 25 g
Product Details & Documents
  • CAS Number:3034-53-5
  • Formula:C₃H₂BrNS
  • MDL Number:MFCD00005316
  • Molecular Weight:164.03 g/mol
  • Boiling Pt:171 °C
  • Density:1.82 g/cm³ (25 °C)
  • Flash Pt:67 °C
  • Storage Temperature:Refrigerator
  • EINECS:221-229-4
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    25 g

Specifications