2-Bromothiazole
Bottle, 100 g
Catalog # APOSOR23133-100G
Supplier: Apollo Scientific
Jump to:
Product Details & Documents
- CAS Number:3034-53-5
- Formula:C₃H₂BrNS
- MDL Number:MFCD00005316
- Molecular Weight:164.03 g/mol
- Boiling Pt:171 °C
- Density:1.82 g/cm³ (25 °C)
- Flash Pt:67 °C
- Storage Temperature:Refrigerator
- EINECS:221-229-4
Specifications
Specifications
- Pack type:Bottle
- Pk:100 g
- Cat. No.:APOSOR23133-100G



