Order Entry
Germany
Orders LinkContactUsLinkComponent
Ninhydrin 99% | Avantor

Ninhydrin 99%

Bottle, 25 g
Catalog # APOSOR2351-25G
Irritant
Supplier:   Apollo Scientific
Ninhydrin 99%
Catalog # APOSOR2351-25G
Supplier:  Apollo Scientific
CAS Number:  485-47-2
Ninhydrin 99%

Bottle, 25 g

CAS Number:  485-47-2
Price
30,40 €
Log in for availability
1 * 25 g
Product Details & Documents
Synonyms:  Ninhydrin monohydrate, Trioxohydrindene monohydrate, 2,2-Dihydroxyindane-1,3-dione, 2,2-Dihydroxy-1,3-indanedione, Indanetrione hydrate, 2,2-Dihydroxyindan-1,3-dion, 1,2,3-Indantrione monohydrate
  • CAS Number:485-47-2
  • Formula:C₉H₆O₄
  • MDL Number:MFCD00003791
  • Molecular Weight:178.14 g/mol
  • Melting Pt:250 °C (dec.)
  • Storage Temperature:Ambient
  • EINECS:207-618-1

Documents

Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    25 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.