2-(2,2-Difluoroethoxy)ethanol
Bottle, 5 g
Catalog # APOSPC450198-5G
Supplier: Apollo Scientific
2-(2,2-Difluoroethoxy)ethanol
Catalog # APOSPC450198-5G
Supplier: Apollo Scientific
CAS Number: 853005-07-9
Jump to:
Product Details & Documents
- CAS Number:853005-07-9
- Formula:C₄H₈F₂O₂
- MDL Number:MFCD19442036
- Molecular Weight:126.102 g/mol
- Storage Temperature:Refrigerator
- UN:1993
- ADR:3,III
Specifications
Specifications
- Pack type:Bottle
- Pk:5 g
- Cat. No.:APOSPC450198-5G
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



