Order Entry
Germany
Orders LinkContactUsLinkComponent
2-(2,2-Difluoroethoxy)ethanol

2-(2,2-Difluoroethoxy)ethanol

Bottle, 25 g
Catalog # APOSPC450198-25G
Flammable
Irritant
Supplier:   Apollo Scientific
2-(2,2-Difluoroethoxy)ethanol
Catalog # APOSPC450198-25G
Supplier:  Apollo Scientific
CAS Number:  853005-07-9
2-(2,2-Difluoroethoxy)ethanol

Bottle, 25 g

CAS Number:  853005-07-9
Price
956,00 €
Log in for availability
1 * 25 g
Product Details & Documents
  • CAS Number:853005-07-9
  • Formula:C₄H₈F₂O₂
  • MDL Number:MFCD19442036
  • Molecular Weight:126.102 g/mol
  • Storage Temperature:Refrigerator
  • UN:1993
  • ADR:3,III
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    25 g

Specifications