Order Entry
Germany
Orders LinkContactUsLinkComponent
2-Hydroxyisonicotinic acid

2-Hydroxyisonicotinic acid

Bottle, 25 g
Catalog # APOSOR1910-25G
Irritant
Supplier:   Apollo Scientific
2-Hydroxyisonicotinic acid
Catalog # APOSOR1910-25G
Supplier:  Apollo Scientific
CAS Number:  22282-72-0
2-Hydroxyisonicotinic acid

Bottle, 25 g

CAS Number:  22282-72-0
Price
46,40 €
Log in for availability
1 * 25 g
Product Details & Documents
Synonyms:  2-Hydroxy-4-pyridinecarboxylic acid, 2-Hydroxy-isonicotinic acid
  • CAS Number:22282-72-0
  • Formula:C₆H₅NO₃
  • MDL Number:MFCD01318566
  • Molecular Weight:139.11 g/mol
  • Melting Pt:>300 °C
  • Melting Pt:>300 °C
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    25 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.