Order Entry
Germany
Orders LinkContactUsLinkComponent
2-Hydroxyisonicotinic acid

2-Hydroxyisonicotinic acid

Bottle, 100 g
Catalog # APOSOR1910-100G
Irritant
Supplier:   Apollo Scientific
2-Hydroxyisonicotinic acid
Catalog # APOSOR1910-100G
Supplier:  Apollo Scientific
CAS Number:  22282-72-0
2-Hydroxyisonicotinic acid

Bottle, 100 g

CAS Number:  22282-72-0
Price
160,00 €
Log in for availability
1 * 100 g
Product Details & Documents
Synonyms:  2-Hydroxy-4-pyridinecarboxylic acid, 2-Hydroxy-isonicotinic acid
  • CAS Number:22282-72-0
  • Formula:C₆H₅NO₃
  • MDL Number:MFCD01318566
  • Molecular Weight:139.11 g/mol
  • Melting Pt:>300 °C
  • Melting Pt:>300 °C
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 g

Specifications