2-(Methylthio)nicotinoyl chloride
Bottle, 1 g
Catalog # APOSOR21399-1G
Supplier: Apollo Scientific
2-(Methylthio)nicotinoyl chloride
Catalog # APOSOR21399-1G
Supplier: Apollo Scientific
CAS Number: 97936-43-1
Jump to:
Product Details & Documents
- CAS Number:97936-43-1
- Formula:C₇H₆ClNOS
- MDL Number:MFCD00051678
- Molecular Weight:187.65 g/mol
- Melting Pt:92…96 °C
- Storage Temperature:Ambient
- UN:3261
- ADR:8,II
Specifications
Specifications
- Pack type:Bottle
- Pk:1 g



