4'-Propylacetophenone
Bottle, 50 g
Catalog # APOSOR1514-50G
Supplier: Apollo Scientific
Jump to:
Product Details & Documents
Synonyms: 1-(4-Propylphenyl)ethanone, p-Propylacetophenone
- CAS Number:2932-65-2
- Formula:C₁₁H₁₄O
- MDL Number:MFCD00041359
- Molecular Weight:162.23
- Boiling Pt:123…125 °C/15mmHg
- Storage Temperature:Ambient
Specifications
Specifications
- Pack type:Bottle
- Pk:50 g



