Order Entry
Germany
Orders LinkContactUsLinkComponent
 

4'-Propylacetophenone

Bottle, 10 g
Catalog # APOSOR1514-10G
Irritant
Supplier:   Apollo Scientific
4'-Propylacetophenone
Catalog # APOSOR1514-10G
Supplier:  Apollo Scientific
CAS Number:  2932-65-2
4'-Propylacetophenone

Bottle, 10 g

CAS Number:  2932-65-2
Price
21,80 €
Log in for availability
1 * 10 g
Product Details & Documents
Synonyms:  1-(4-Propylphenyl)ethanone, p-Propylacetophenone
  • CAS Number:2932-65-2
  • Formula:C₁₁H₁₄O
  • MDL Number:MFCD00041359
  • Molecular Weight:162.23
  • Boiling Pt:123…125 °C/15mmHg
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    10 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.