2-Fluorobutane
Bottle, 5 g
Catalog # APOSPC408041-5G
Supplier: Apollo Scientific
Jump to:
Product Details & Documents
- CAS Number:359-01-3
- Formula:C₄H₉F
- MDL Number:MFCD02258657
- Molecular Weight:76.113 g/mol
- Boiling Pt:25 °C
- Melting Pt:–121 °C
- Density:0.77 g/cm³ (25 °C)
- Storage Temperature:Refrigerator
- UN:1993
- ADR:3,I
Specifications
Specifications
- Pack type:Bottle
- Pk:5 g



