Order Entry
Germany
Orders LinkContactUsLinkComponent
 

4-Fluorophenol 99%

Bottle, 100 g
Catalog # APOSPC4090-100G
Corrosive
Toxic
Supplier:   Apollo Scientific
4-Fluorophenol 99%
Catalog # APOSPC4090-100G
Supplier:  Apollo Scientific
CAS Number:  371-41-5
4-Fluorophenol 99%

Bottle, 100 g

CAS Number:  371-41-5
Price
23,20 €
Log in for availability
1 * 100 g
Product Details & Documents
Synonyms:  p-Fluorophenol, 1-Fluoro-4-hydroxybenzene
  • CAS Number:371-41-5
  • Formula:C₆H₅FO
  • MDL Number:MFCD00002316
  • Molecular Weight:112.1 g/mol
  • Boiling Pt:185 °C
  • Melting Pt:44…47 °C
  • Density:1.22 g/cm³ (25 °C)
  • Flash Pt:68 °C
  • Storage Temperature:Ambient
  • EINECS:206-736-0
  • UN:2928
  • ADR:6.1+8,II
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 g
  • Cat. No.:
    APOSPC4090-100G

Specifications