Order Entry
Germany
Orders LinkContactUsLinkComponent
 

3-Fluoropyrocatechol 99%

Bottle, 5 g
Catalog # APOSPC3728D-5G
Ätzend
Reizend
Supplier:   Apollo Scientific
3-Fluoropyrocatechol 99%
Catalog # APOSPC3728D-5G
Supplier:  Apollo Scientific
CAS Number:  363-52-0
3-Fluoropyrocatechol 99%

Bottle, 5 g

Wechseln zu:
CAS Number:  363-52-0
Price
39,10 €
Anmelden für die Verfügbarkeit
1 * 5 g
Product Details & Documents
Synonyme:  3-Fluorocatechol, 3-Fluorobenzene-1,2-diol, 3-Fluoro-1,2-dihydroxybenzene
  • CAS-Nummer:363-52-0
  • Formel::C₆H₅FO₂
  • MDL-Nummer:MFCD00042582
  • Molekulargewicht:128.1 g/mol
  • Schmelzpunkt:71…73 °C
  • Lagertemperatur:Ambient
  • UN:3261
  • ADR:8,II
Spezifikationen:

Spezifikationen:

  • Pack type:
    Bottle
  • Pk:
    5 g

Spezifikationen:

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.