Order Entry
Germany
Orders LinkContactUsLinkComponent
3-Fluoropyrocatechol 99%

3-Fluoropyrocatechol 99%

Bottle, 1 g
Catalog # APOSPC3728D-1G
Corrosive
Irritant
Supplier:   Apollo Scientific
3-Fluoropyrocatechol 99%
Catalog # APOSPC3728D-1G
Supplier:  Apollo Scientific
CAS Number:  363-52-0
3-Fluoropyrocatechol 99%

Bottle, 1 g

CAS Number:  363-52-0
Price
21,80 €
Log in for availability
1 * 1 g
Product Details & Documents
Synonyms:  3-Fluorocatechol, 3-Fluorobenzene-1,2-diol, 3-Fluoro-1,2-dihydroxybenzene
  • CAS Number:363-52-0
  • Formula:C₆H₅FO₂
  • MDL Number:MFCD00042582
  • Molecular Weight:128.1 g/mol
  • Melting Pt:71…73 °C
  • Storage Temperature:Ambient
  • UN:3261
  • ADR:8,II
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.