Order Entry
Germany
Orders LinkContactUsLinkComponent
Xanthine

Xanthine

Bottle, 25 g
Catalog # APOSBIX1002-25G
Irritant
Supplier:   Apollo Scientific
Xanthine
Catalog # APOSBIX1002-25G
Supplier:  Apollo Scientific
CAS Number:  69-89-6
Xanthine

Bottle, 25 g

CAS Number:  69-89-6
Price
29,00 €
Log in for availability
1 * 25 g
Product Details & Documents
Synonyms:  1H-Purine-2,6(3H,7H)-dione, 3,7-Dihydro-1H-purine-2,6-dione, 2,6-Dihydroxypurine
  • CAS Number:69-89-6
  • Formula:C₅H₄N₄O₂
  • MDL Number:MFCD00078453
  • Molecular Weight:152.11 g/mol
  • Melting Pt:300 °C
  • Storage Temperature:Ambient
  • EINECS:200-718-6

Documents

Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    25 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.