Order Entry
Germany
Orders LinkContactUsLinkComponent
Valinomycin

Valinomycin

Bottle, 25 mg
Catalog # APOSBIV0893-25MG
Toxic
Supplier:   Apollo Scientific
Valinomycin
Catalog # APOSBIV0893-25MG
Supplier:  Apollo Scientific
CAS Number:  2001-95-8
Valinomycin

Bottle, 25 mg

CAS Number:  2001-95-8
Price
65,20 €
Log in for availability
1 * 25 mg
Product Details & Documents

Anitbiotic produced by Streptomyces fulvissimus. Valinomycin is highly selective for K+ ions over Na+without influencing the permeability of other ions

  • CAS Number:2001-95-8
  • Formula:C₅₄H₉₀O₁₈N₆
  • MDL Number:MFCD00005114
  • Molecular Weight:1111.34 g/mol
  • Melting Pt:187…190 °C
  • Storage Temperature:Refrigerator
  • EINECS:217-896-6
  • UN:2811
  • ADR:6.1,I
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    25 mg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.