Order Entry
Germany
Orders LinkContactUsLinkComponent
 

Vancomycin hydrochloride

Bottle, 5 g
Catalog # APOSBIV0155-5G
Irritant
Supplier:   Apollo Scientific
Vancomycin hydrochloride
Catalog # APOSBIV0155-5G
Supplier:  Apollo Scientific
CAS Number:  1404-93-9
Vancomycin hydrochloride

Bottle, 5 g

CAS Number:  1404-93-9
Price
110,00 €
Log in for availability
1 * 5 g
Product Details & Documents

Vancomycin is a glycopeptide antibiotic. It exerts its action by inhibiting the formation of the peptidoglycan polymers of the bacterial cell wall. Often used incombination with cefotaxim or carbenicillin.

  • Potency of 1099 mcg/mg
  • Cell culture tested.
Synonyms:  (3S,6R,7R,22R,23S,26S,36R,38aR)-3-(2-Amino-2-oxoethyl)-44-[[2-O-(3-amino-2,3,6-trideoxy-3-C-methyl-α-L-lyxo-hexopyranosyl)-β-D-glucopyranosyl]oxy]-10,19-dichloro-2,3,4,5,6,7,23,24,25,26,36,37,38,38a-tetradecahydro-7,22,28,30,32-pentahydroxy-6-[[(2R)-4-methyl-2-(methylamino)-1-oxopentyl]amino]-2,5,24,38,39-pentaoxo-22H-8,11:18,21-Dietheno-23,36-(iminomethano)-13,16:31,35-dimetheno-1H,16H-[1,6,9]oxadiazacyclohexadecino[4,5-m][10,2,16]benzoxadiazacyclotetracosine-26-carboxylic acid xhydrochloride, Vancomycin supplement
  • CAS Number:1404-93-9
  • Formula:C₆₆H₇₅Cl₂N₉O₂₄·HCl
  • MDL Number:MFCD03613611
  • Molecular Weight:1485.71 g/mol
  • Storage Temperature:Refrigerator
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g

Specifications