Order Entry
Germany
Orders LinkContactUsLinkComponent
Tobramycin

Tobramycin

Bottle, 1 g
Catalog # APOSBIT0802-1G
Irritant
Supplier:   Apollo Scientific
Tobramycin
Catalog # APOSBIT0802-1G
Supplier:  Apollo Scientific
CAS Number:  32986-56-4
Tobramycin

Bottle, 1 g

CAS Number:  32986-56-4
Price
203,00 €
Log in for availability
1 * 1 g
Product Details & Documents

Tobramycin is an aminoglycoside antibiotic and hasa bactericidal action against many gram-negative bacteria. Aminoglycosides are taken into sensitivebacterial cells by an active transport process.

Synonyms:  (2S,3R,4S,5S,6R)-4-Amino-2-(((1S,2S,3R,4S,6R)-4,6-diamino-3-(((2R,3R,5S,6R)-3-amino-6-(aminomethyl)-5-hydroxytetrahydro-2H-pyran-2-yl)oxy)-2-hydroxycyclohexyl)oxy)-6-(hydroxymethyl)tetrahydro-2H-pyran-3,5-diol
  • CAS Number:32986-56-4
  • Formula:C₁₈H₃₇N₅O₉
  • MDL Number:MFCD00077885
  • Molecular Weight:467.51 g/mol
  • Storage Temperature:Refrigerator
  • EINECS:251-322-5
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.