Order Entry
Germany
Orders LinkContactUsLinkComponent
2-Chloro-6-nitrophenol

2-Chloro-6-nitrophenol

Bottle, 5 g
Catalog # APOSOR400451-5G
Irritant
Supplier:   Apollo Scientific
2-Chloro-6-nitrophenol
Catalog # APOSOR400451-5G
Supplier:  Apollo Scientific
CAS Number:  603-86-1
2-Chloro-6-nitrophenol

Bottle, 5 g

CAS Number:  603-86-1
Price
24,60 €
Log in for availability
1 * 5 g
Product Details & Documents
  • CAS Number:603-86-1
  • Formula:C₆H₄ClNO₃
  • MDL Number:MFCD00024233
  • Molecular Weight:173.56 g/mol
  • Melting Pt:67…70 °C
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g

Specifications