Order Entry
Germany
Orders LinkContactUsLinkComponent
β-Cyclodextrine

β-Cyclodextrine

Bottle, 500 g
Catalog # APOSBIM1015-500G
Irritant
Supplier:   Apollo Scientific
β-Cyclodextrine
Catalog # APOSBIM1015-500G
Supplier:  Apollo Scientific
CAS Number:  7585-39-9
β-Cyclodextrine

Bottle, 500 g

CAS Number:  7585-39-9
Price
261,00 €
Log in for availability
1 * 500 g
Product Details & Documents
Synonyms:  Caraway, Cycloheptaamylose, β-Cyclodextrin, Cyclomaltoheptaose, Schardinger β-Dextrin, Betadex
  • CAS Number:7585-39-9
  • Formula:C₄₂H₇₀O₃₅
  • MDL Number:MFCD00078139
  • Molecular Weight:1134.98 g/mol
  • Melting Pt:290…300 °C
  • Density:1.44 g/cm³ (25 °C)
  • Storage Temperature:Ambient

Documents

Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    500 g

Specifications