Order Entry
Germany
Orders LinkContactUsLinkComponent
Digitonin

Digitonin

Bottle, 1 g
Catalog # APOSBID3301-1G
Toxic
Supplier:   Apollo Scientific
Digitonin
Catalog # APOSBID3301-1G
Supplier:  Apollo Scientific
CAS Number:  11024-24-1
Digitonin

Bottle, 1 g

CAS Number:  11024-24-1
Price
160,00 €
Log in for availability
1 * 1 g
Product Details & Documents

Non-ionic detergent used to permeabilize cell membranes.

Synonyms:  Digitonin
  • CAS Number:11024-24-1
  • Formula:C₅₆H₉₂O₂₉
  • MDL Number:MFCD00077729
  • Molecular Weight:1229.312 g/mol
  • Storage Temperature:Ambient
  • EINECS:234-255-6
  • UN:3462
  • ADR:6.1,III

Documents

Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g

Specifications