Auftragserfassung
Germany
AuftragsverfolgungKontakt
Bromphenolblau

Bromphenolblau

Bottle, 10 g
Catalog # APOSBIB1005-10G
Irritant
Supplier:   Apollo Scientific
Bromphenolblau
Catalog # APOSBIB1005-10G
Supplier:  Apollo Scientific
CAS Number:  115-39-9
Bromphenolblau

Bottle, 10 g

CAS Number:  115-39-9
Price
55,00 €
Log in for availability
1 * 10 g
Product Details & Documents
Synonyms:  3,3',5,5'-Tetrabromphenolsulfonphthalein, 4,4'-(3H-2,1-Benzoxathiol-3-yliden)bis[2,6-dibromphenol]-S,S-dioxid
  • CAS Number:115-39-9
  • Formula:C₁₉H₁₀Br₄O₅S
  • MDL Number:MFCD00005875
  • Molecular Weight:669.97 g/mol
  • Melting Pt:279 °C
  • Storage Temperature:Raumtemperatur

Documents

Specifications

Specifications

  • Packung:
    Bottle
  • VE:
    10 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.