L-Methionine ≥98.5% USP
Glass, amber, 500 g
Supplier: Spectrum Chemical
Jump to:
Product Details & Documents
Methionine, USP is used to prevent liver damage in acetaminophen poisoning. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Synonyms: (S)-(-)-2-Amino-4-(methylthio)butyric acid, (S)-(-)-Methionine, L(-)-Methionine, (S)-(-)-2-Amino-4-(methylthio)butanoic acid, H-Met-OH, Methionine, (S)-2-Amino-4-(methylthio)butanoic acid
- CAS Number:63-68-3
- Formula:C₅H₁₁NO₂S
- MDL Number:MFCD00063097
- Molecular Weight:149.21 g/mol
- Storage Temperature:Ambient
Documents
Certificates
Search by Lot NumberSpecifications
Specification Test Results
- Assay (Dried Basis):98.5 - 101.5%
- Specific Rotation:+22.4 to +24.7°
- pH (1 in 100):5.6 - 6.1
- Loss On Drying:max. 0.3 %
- Related Compounds:To Pass Test
Specifications
- Pack type:Glass, amber
- Pk:500 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



