Robotic tips (CO-RE Tips) and consumables for Microlab
Product Details & Documents
Hamilton CO-RE tips were designed for MICROLAB pipetting workstations in order to solve the problem of sealing and alignment plagued by press-fit tip attachment techniques. Each CO-RE disposable tip has a circular groove inside the attachment orifice. An o-ring on the pipetting channel is expanded to fit this groove precisely providing an air-tight seal and straight alignment between tip and pipetting channel.
- Pressure-based Liquid Level Detection (pLLD) and capacitive LLD (cLLD) with conductive (black) version
- DNase-/RNase-free, pyrogen-free (endotoxin-free), PCR inhibitor-free
- Stackable tip racks increase tip capacity on the workstation deck for longer walk away times: 1920 tips can be loaded in one step
- Racks allow tips to be picked up by the independent channels or the 96-channel multiprobe head directly from the stack
The superior biological purity of the tips is guaranteed by a controlled production process. They are therefore suitable for both diagnostic and pharmaceutical laboratories in Genomics, Proteomics, Cellomics, Forensic and drug discovery fields.
Filter tips
- Maximum consistency in air flow and chosen porosity of filter element ensure optimal pipetting and dispensing performance across a wide range of liquid volumes
- Bacterial aerosol filtration efficiency of >99,9%
SLIM tips
- Minimise risk of liquid overflow in cluster tubes, deep-well blocks and two-phased liquid applications
- Especially for aspirating liquid in 'fixed height mode' down to the base of thin tubes
- Slim tips require labware files, liquid classes for single channels and CO-RE 96 probe head
Rocket tips
- 4-to-1 adapter enables flexible pipetting and saves times: No need for a second head or a complex head exchange
- For pipetting in 96 deep-well blocks and microtiter plates with well diameters ≥3,7 mm
Wide bore tips
- CO-RE tips with various distal end orifices which provide the flexibility when handling difficult samples
- Typical application areas: genomic DNA, fragile cell lines, blood fractionation to minimise shearing forces, pipetting beads or other viscosity liquids
Piercing tips
- Optimised to aspirate and dispense into various sealed labware which is pierceable with ≤10 N
- Optimum material composition – carbon fibre reinforcement allows piercing of various sealing materials
SLIM tips
- Minimise risk of liquid overflow in cluster tubes, deep-well blocks and two-phased liquid applications
- Especially for aspirating liquid in “fixed height mode” down to the base of thin tubes
- 40% reduction of displaced liquid volume, compared to the CO-RE 1000 µl tip
- Slim tips require labware files, liquid classes for single channels and CO-RE 96 probe head
Rocket tips
- Unique Rocket tip design with 4-to-1 adapter: Usable on the CO-RE 384 probe head
- Adapter enables flexible pipetting and saves times: No need for a second head or a complex head exchange
- For pipetting in 96 deep-well blocks and microtiter plates with well diameters ≥ 3,7 mm
Accessories information: Reagent containers for Microlab STAR line
- Made of PP, autoclavable, with 60 or 120 ml capacity
- Can be placed in a reagent carrier on STAR line instruments
- With volume markings, without lid, with one (60 ml) or three (120 ml) wave breakers to avoid splashes during movements of the filled container
Deep-well blocks for automation
- Ideal for automated processing or handling large quantities of backup samples
- PP material is ideal for backup storage of pharmaceutical substances, DNA isolation, long-term sample storage, enzyme and tissue culture applications
- PS blocks are ideal for long-term storage of blood samples; easy optical control due to transparent wells
Waste bags, liquid-tight, welded
- Disposable PE bags (100 µm thick) are ideal for the containment and disposal of used consumables
- Bags fit on the waste hoop of the STAR line
Waste container and chutes for hazardous waste
- Container for hazardous waste is made of white cardboard, incl. yellow PE bag
- PE waste chutes are open and perforated on both sides, must be used with waste container
Packaging: Tips are either supplied in stackable Nested Tip Rack (NTR) packs – containing a disposable frame and five stacks with four tip racks – or in a blister pack containing five preloaded trays in a clear, recyclable PET thermoformed tray with a sealed paper lid.
Dokumente
Certificates
Search by Lot NumberHAMI235904_TDS_EN HAMI235939_TDS_EN HAMI235948_TDS_EN
Technisches Datenblatt
Specifications
- Packed:NTR pack (10 × 96 tips)
- Sterile:+
- Volume:0,5 - 50 µl
- Length:50 mm
- Seq:18
- Tip style:Low volume, conductive
- Tip colour:Black
- Filter:+
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



