Orders LinkContactUsLinkComponent
Decontamination solution, DNA AWAY®
Decontamination solution, DNA AWAY®
DNA AWAY®, 4 L
Catalog # 732-4436
Decontamination solution, DNA AWAY®
Catalog # 732-4436
CAS Number:  
Decontamination solution, DNA AWAY®
Decontamination solution, DNA AWAY®
DNA AWAY®, 4 L
CAS Number:  
Price
101,00 €
Sale
Valid until 30/06/2026
Log in for availability
1 * 4 L
Product Details & Documents
Ready-to-use solution for eliminating DNA contamination. For cleaning work surfaces, instruments, pipettes, thermal cyclers and more.
  • Suitable for PCR applications, preventing false-positive PCR results
  • Easy cleaning of various types of surfaces
Specifications
Specifications
  • Pk:
    4 L
  • Description:
    DNA AWAY®