Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Fe in Salpetersaure 0.5 mol/l ruckfuhrbar auf NIST 1 * 250 mL
Catalog # KRAF03840.1000
Supplier:
Bernd Kraft
Expand image
Fe in Salpetersaure 0.5 mol/l ruckfuhrbar auf NIST 1 * 250 mL
Catalog # KRAF03840.1000
Supplier:
Bernd Kraft