Supelcarb(R) HC Hydrocarbon Trap, volume 750 cc, fitting size 1/2 in. 1 * 1 items
Catalog # 24565.
Supplier: Merck
Supelcarb(R) HC Hydrocarbon Trap, volume 750 cc, fitting size 1/2 in. 1 * 1 items
Catalog # 24565.
Supplier: Merck
Frequently Bought Together
VWR®, Vial Racks
Supplier: VWR Collection
VWR®, Storage Boxes for Sample Vials and Bottles, PP
Supplier: VWR Collection
TLC plates, silica gel 60G, glass backed
Supplier: Merck
VWR®, Screw Caps for Short Thread Vials ND9
Supplier: VWR Collection
VWR®, Short Thread Vials ND9
Supplier: VWR Collection
Precision screw caps, 18 mm, level 3 high performance applications, SureSTART™
Supplier: Thermo Fisher Scientific
Bonded screw caps for screw neck vials, N 15 (15-425)
Supplier: MACHEREY-NAGEL
Florisil® 60-100 mesh for chromatography
Supplier: VWR Chemicals
Syringes for GC Agilent autosampler 7673-7683, 6850
Supplier: HAMILTON BONADUZ
VWR®, Crimp Neck Vials ND11
Supplier: VWR Collection
VWR®, Snap Cap Vials
Supplier: VWR Collection
Calibration standard, single element, silicon for AAS and ICP-OES, pure grade
Supplier: PerkinElmer
Headspace crimp caps with 20 mm septa
Supplier: Agilent