Supelcarb(R) HC Hydrocarbon Trap, volume 120 cc, fitting size 1/4 in. 1 * 1 items
Catalog # 24449.
Supplier: Merck
Supelcarb(R) HC Hydrocarbon Trap, volume 120 cc, fitting size 1/4 in. 1 * 1 items
Catalog # 24449.
Supplier: Merck
Frequently Bought Together
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Florisil® 60-100 mesh for chromatography
Supplier: VWR Chemicals
VWR®, Crimp Neck Vials ND11
Supplier: VWR Collection
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
VWR®, Short Thread Vials ND9
Supplier: VWR Collection
VWR®, Screw Neck Headspace Vials ND18
Supplier: VWR Collection
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
VWR®, Crimp Caps for Crimp Neck Vials ND11
Supplier: VWR Collection
Liquid-liquid extraction, EXtrelut® NT
Supplier: Merck
VWR®, Snap Ring Vials ND11
Supplier: VWR Collection
VWR®, Snap Cap Vials
Supplier: VWR Collection
Exhaust filters, V3.0
Supplier: S.C.A.T.
Headspace crimp caps with 20 mm septa
Supplier: Agilent