96-Well plate sample control system, U-2D™
Product Details & Documents
The U-2D™ universal two dimensional 96-well plate system offer users unprecedented sample management control compared to current glass lined or plastic well plates for sample preparation or chromatography.
Easily push up the insert from the bottom and remove it from the rack. These inserts can then be placed into any MicroSolv 9 mm screw top autosampler vial for safe storage, transport or for a second chromatography dimension, or to another instrument platform.
Ordering information: Base and sealing mats sold separately.
Specifications
- Description:U-2D™ rack with 96 well plate, 1 ml preloaded inserts, base and plug-style non-slit silicone sealing mat
- Seq:07
Accessories
Product Information
- Catalog #AvailabilityPrice
- Paddles for U-2D™ sealing film
Product Information
- Catalog #AvailabilityPrice
- Roller for U-2D™ sealing film
Product Information
- Catalog #AvailabilityPrice
- U-2D™ plate, 350µL, with rack and base onlyU-2D™ plate, 350µL, with rack and base onlyMICS95296-U2DLog in for availability1 * 1 ST - 116,00 €
Product Information
- Catalog #AvailabilityPrice
- Zone free sealing film for U-2D™ platesZone free sealing film for U-2D™ platesMICS95296-ZFF-PKLog in for availability1 * 10 ST - 54,50 €
Product Information
- Catalog #AvailabilityPrice
- Zone free sealing film for U-2D™ platesZone free sealing film for U-2D™ platesMICS95296-ZFF-CSLog in for availability1 * 50 ST - 228,00 €