ContactUsLinkComponent
Mouse TIGIT, His Tag (TIT-M52E6) is expressed from human 293 cells (HEK293). It contains AA Gly 26 - Thr 143 (Accession NP001139797. 1). 1 * 50 µG
Catalog # ACRBTIT-M52E6-50UG
Supplier:  ACRO Biosystems
Mouse TIGIT, His Tag (TIT-M52E6) is expressed from human 293 cells (HEK293). It contains AA Gly 26 - Thr 143 (Accession NP001139797. 1). 1 * 50 µG
Catalog # ACRBTIT-M52E6-50UG
Supplier:  ACRO Biosystems
Frequently Bought Together