HIGH TEMP. EL FILAMENT FOR AGILENT GC-MS 1 * 1 ST
Catalog # REST23099
Supplier: Restek
Frequently Bought Together
VWR®, Crimp Neck Vials ND11
Supplier: VWR Collection
Methanol ≥99.8%, HiPerSolv CHROMANORM®, Isocratic Grade for HPLC
Supplier: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Acetone ≥99.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
VWR®, Short Thread Vials ND9
Supplier: VWR Collection
VWR®, Screw Neck Headspace Vials ND18
Supplier: VWR Collection
Acetone ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Pasteur pipettes
Supplier: HIRSCHMANN
Exhaust filters, V3.0
Supplier: S.C.A.T.
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
VWR®, Nitrile Gloves
Supplier: VWR Collection