Order Entry
Germany
Orders LinkContactUsLinkComponent
Containment berms, QuickBerm® Lite

Containment berms, QuickBerm® Lite

Rigid-lock QUICKBERM® lite, folded dimensions (279×203×127 mm), 4,5 kg, 299 l, 1200×1200×203 mm
Catalog # 119-0373
Containment berms, QuickBerm® Lite
Catalog # 119-0373
Containment berms, QuickBerm® Lite
Containment berms, QuickBerm® Lite

Rigid-lock QUICKBERM® lite, folded dimensions (279×203×127 mm), 4,5 kg, 299 l, 1200×1200×203 mm

Price
667,00 €
Log in for availability
1 * 1 ST
Product Details & Documents

Portable, versatile containment berm deploys quickly for emergency spill response.

  • Leakproof, tough Durable EnGuard™I, PVC coated fabric provides broad UV and chemical resistance that withstands repeated usage
  • Reinforced inside supports reduce sagging and prevent tripping hazards that can be caused by outside supports

Lightweight berm folds down easily to a small size for storage in trucks, emergency vehicles, and spill kits. Add optional grates to help reduce contact with contaminated liquids by personnel or equipment.

Certifications: Helps to comply with EPA for containment and spill prevention (SPCC).

Specifications
  • Capacity:
    299 l
  • Description:
    Rigid-lock QUICKBERM® lite, folded dimensions (279×203×127 mm), 4,5 kg
  • Seq:
    01
  • W×D×H:
    1200×1200×203 mm