Drip pads
Product Details & Documents
Drip pads can capture small oil leaks and drips before they contaminate the ground or create unsafe work surfaces.
- Lightweight, flexible, and portable
- Made from UV and chemical-resistant modified PVC
- Coated mesh outer layer to trap coarse debris
These drip pads feature replaceable, oil-only sorbent with a welded leakproof barrier. Rated for drive over applications, pads include brass grommets in opposing corners for anchoring. Anchoring stakes not included. Pads come in two sizes; extended model is weighted to keep it down in high winds. Outer shell is reusable with optional refill sorbent packs.
Certifications: Helps to comply with EPA for spill prevention (SPCC).
Specifications
- Capacity:7 l
- Description:Pad drip refills
- L×W:1321 × 762 mm
- Seq:04
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



